![Anti-KMT2E antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/269/122/6be9d379-c862-40b6-90c7-ce8af97c77d9/640/6be9d379-c862-40b6-90c7-ce8af97c77d9.jpg)
Anti-KMT2E antibody produced in rabbit
![Anti-KMT2E antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/269/122/6be9d379-c862-40b6-90c7-ce8af97c77d9/640/6be9d379-c862-40b6-90c7-ce8af97c77d9.jpg)
Synonym(s):
Anti-HDCMC04P, Anti-MLL5
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing