Anti-HDAC1 antibody produced in rabbit
Synonym(s):
Anti-GON-10, Anti-HD1, Anti-RPD3L1, Anti-histone deacetylase 1, HD1, Histone deacetylase 1, RPD3L1, Anti-DKFZp686H12203, Anti-Histone deacetylase 1, Anti-RPD3
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing