Anti-Ganglioside GM₁ Rabbit pAb
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Post Translational Modification
Species Reactivity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing