
Anti-EPO-R antibody produced in rabbit

Synonym(s):
EPO-R, EPOR, Erythropoietin receptor precursor, Anti-Erythropoietin receptor, Anti-MGC138358
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing