
Anti-CSAD antibody produced in rabbit

Synonym(s):
Anti-CSD, Anti-Cysteine sulfinic acid decarboxylase, Anti-PCAP
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing