Skip to Content
MilliporeSigma
Acetonitrile-1-13C,15N 99 atom % 13C, 98 atom % 15N

Acetonitrile-1-13C,15N

Acetonitrile-1-13C,15N 99 atom % 13C, 98 atom % 15N
Synonym(s):
Methyl cyanide-13C,15N
Linear Formula:
CH313C15N
CAS Number:
Molecular Weight:
43.04

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing