![8-Chloro-2-ethylimidazol[1,2-a]pyrazine](/deepweb/assets/sigmaaldrich/product/structures/253/432/d8c96ae7-c804-40ef-a6ae-624e7868c516/640/d8c96ae7-c804-40ef-a6ae-624e7868c516.png)
8-Chloro-2-ethylimidazol[1,2-a]pyrazine
![8-Chloro-2-ethylimidazol[1,2-a]pyrazine](/deepweb/assets/sigmaaldrich/product/structures/253/432/d8c96ae7-c804-40ef-a6ae-624e7868c516/640/d8c96ae7-c804-40ef-a6ae-624e7868c516.png)
Empirical Formula (Hill Notation):
C8H8ClN3
CAS Number:
Molecular Weight:
181.62
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing