Skip to Content
MilliporeSigma
6-tert-Butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-hydroxy-2,4-dimethyl-cyclohex-2,5-dienone trifluoroacetate Pharmaceutical Secondary Standard; Certified Reference Material

6-tert-Butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-hydroxy-2,4-dimethyl-cyclohex-2,5-dienone trifluoroacetate

6-tert-Butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-hydroxy-2,4-dimethyl-cyclohex-2,5-dienone trifluoroacetate Pharmaceutical Secondary Standard; Certified Reference Material
Empirical Formula (Hill Notation):
C16H25N2O2 C2F3O2
Molecular Weight:
390.40

Products

Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1