![6-Methoxy-2-naphthoic acid pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/274/639/d4a159e2-feda-44ce-b3ad-9c690c7c08f7/640/d4a159e2-feda-44ce-b3ad-9c690c7c08f7.png)
6-Methoxy-2-naphthoic acid
![6-Methoxy-2-naphthoic acid pharmaceutical impurity standard](/deepweb/assets/sigmaaldrich/product/structures/274/639/d4a159e2-feda-44ce-b3ad-9c690c7c08f7/640/d4a159e2-feda-44ce-b3ad-9c690c7c08f7.png)
Synonym(s):
6-Methoxy-2-naphthalenecarboxylic acid
Linear Formula:
CH3OC10H6CO2H
CAS Number:
Molecular Weight:
202.21
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing