![6-Chloro[1,2,4]triazolo[4,3-b]pyridazine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/410/755/848880cb-3ee5-4463-9046-08b56d8184a0/640/848880cb-3ee5-4463-9046-08b56d8184a0.png)
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
![6-Chloro[1,2,4]triazolo[4,3-b]pyridazine AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/410/755/848880cb-3ee5-4463-9046-08b56d8184a0/640/848880cb-3ee5-4463-9046-08b56d8184a0.png)
Empirical Formula (Hill Notation):
C5H3ClN4
CAS Number:
Molecular Weight:
154.56
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing