
6,7-Dimethylquinoline-2,3-dicarboxylic acid

Empirical Formula (Hill Notation):
C13H11NO4
CAS Number:
Molecular Weight:
245.23
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing