5-Formyl-2-methoxybenzenesulfonamide
Empirical Formula (Hill Notation):
C8H9NO4S
CAS Number:
Molecular Weight:
215.23
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing