
5-Bromo-2-fluoropyrimidine

Empirical Formula (Hill Notation):
C4H2BrFN2
CAS Number:
Molecular Weight:
176.97
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing