
5-(2-Methoxyphenyl)cyclohexane-1,3-dione

Empirical Formula (Hill Notation):
C13H14O3
CAS Number:
Molecular Weight:
218.25
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing