Skip to Content
MilliporeSigma
(4-Hydroxyphenyl)diphenylphosphine 98%

(4-Hydroxyphenyl)diphenylphosphine

(4-Hydroxyphenyl)diphenylphosphine 98%
Synonym(s):
(p-Hydroxyphenyl)diphenylphosphine
Linear Formula:
(C6H5)2PC6H4OH
CAS Number:
Molecular Weight:
278.28

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing