Skip to Content
MilliporeSigma
4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-α,α-dimethylbenzeneacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-α,α-dimethylbenzeneacetic acid

4-[4-[4-(Diphenylmethylene)-1-piperidinyl]-1-hydroxybutyl]-α,α-dimethylbenzeneacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-[4-[-4-[4-(Diphenylmethylidene)piperidin-1-yl]-1-hydroxybutyl]phenyl]-2-methylpropanoic acid, Fexofenadine Olefin
Empirical Formula (Hill Notation):
C32H37NO3
CAS Number:
Molecular Weight:
483.64

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade