
4-(1H-Indol-3-yl)butan-2-one

Synonym(s):
3-(3-Oxobutyl)-1H-indole
Empirical Formula (Hill Notation):
C12H13NO
CAS Number:
Molecular Weight:
187.24
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1