<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
(3-NITROPHENYL)PHOSPHONIC ACID
Linear Formula:
C
6
H
6
NO
5
P
CAS Number:
5337-19-9
Molecular Weight:
203.092
Products
Formula Weight
Special Grade
Aldrich
CPR
(2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
S915734
(3-NITROPHENYL)PHOSPHONIC ACID,
Aldrich
CPR
Expand
View Pricing
S311529
(3-NITROPHENYL)PHOSPHONIC ACID,
Aldrich
CPR
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
English - EN
Español - ES
Learn More