Skip to Content
MilliporeSigma
3-(Methylthio)hexyl acetate ≥97%, FG

3-(Methylthio)hexyl acetate

3-(Methylthio)hexyl acetate ≥97%, FG
Empirical Formula (Hill Notation):
C9H18O2S
CAS Number:
Molecular Weight:
190.30

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing