Skip to Content
MilliporeSigma
3-Chloro-5-fluorobenzonitrile 97%

3-Chloro-5-fluorobenzonitrile

3-Chloro-5-fluorobenzonitrile 97%
Linear Formula:
ClC6H3(F)CN
CAS Number:
Molecular Weight:
155.56

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing