
3,5-Dinitrocatechol

Synonym(s):
3,5-Dinitro-1,2-benzenediol, OR-486
Empirical Formula (Hill Notation):
C6H4N2O6
CAS Number:
Molecular Weight:
200.11
Products
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing