3,3′-Dimethylnaphthidine
Synonym(s):
3,3′-Dimethylnaphthidine
Empirical Formula (Hill Notation):
C22H20N2
CAS Number:
Molecular Weight:
312.41
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing