![(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/263/882/35c9e494-9f53-45c5-96ab-4f0ca92d7d7d/640/35c9e494-9f53-45c5-96ab-4f0ca92d7d7d.png)
(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic acid
![(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/263/882/35c9e494-9f53-45c5-96ab-4f0ca92d7d7d/640/35c9e494-9f53-45c5-96ab-4f0ca92d7d7d.png)
Synonym(s):
(R)-2-Phenyl-2-pivalamidoacetic acid
Empirical Formula (Hill Notation):
C13H17NO3
CAS Number:
Molecular Weight:
235.28
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1