Skip to Content
MilliporeSigma
2-Methyl-3-tetrahydrofuranthiol mixture of cis and trans, ≥97%, FG

2-Methyl-3-tetrahydrofuranthiol

2-Methyl-3-tetrahydrofuranthiol mixture of cis and trans, ≥97%, FG
Empirical Formula (Hill Notation):
C5H10OS
CAS Number:
Molecular Weight:
118.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing