Skip to Content
MilliporeSigma
2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide Pharmaceutical Secondary Standard; Certified Reference Material

2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide

2-Hydroxy-N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylacetamide Pharmaceutical Secondary Standard; Certified Reference Material
Empirical Formula (Hill Notation):
C11H15NO4
Molecular Weight:
225.24

Products

Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade