
2-Fluoro-3-iodo-5-methylpyridine

Empirical Formula (Hill Notation):
C6H5FIN
CAS Number:
Molecular Weight:
237.01
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing