Skip to Content
MilliporeSigma
2-Ethylfenchol ≥97%, FCC, FG

2-Ethylfenchol

2-Ethylfenchol ≥97%, FCC, FG
Empirical Formula (Hill Notation):
C12H22O
CAS Number:
Molecular Weight:
182.30
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing