Skip to Content
MilliporeSigma
2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane 95%

2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane

2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane 95%
Empirical Formula (Hill Notation):
C6H12ClO2P
CAS Number:
Molecular Weight:
182.59

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing