
2-Chloro-1-(3,4-dimethylphenyl)ethanone

Empirical Formula (Hill Notation):
C10H11ClO
CAS Number:
Molecular Weight:
182.65
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing