Skip to Content
MilliporeSigma
2-amino-4,6-dichlorobenzoic acid AldrichCPR

2-Amino-4,6-dichlorobenzoic acid

2-amino-4,6-dichlorobenzoic acid AldrichCPR
Synonym(s):
2-Amino-4,6-dichlorobenzoic acid
Empirical Formula (Hill Notation):
C7H5Cl2NO2
CAS Number:
Molecular Weight:
206.03

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing