
2,6-Dichloro-N-(2,6-dimethylphenyl)hexanamide

Synonym(s):
Bupivacaine impurity D (PhEur)
Empirical Formula (Hill Notation):
C14H19Cl2NO
CAS Number:
Molecular Weight:
288.21
Products
Brand
Formula Weight
Manufacturer
Product Line
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing