![2-[4-(2-Methylpropyl)phenyl]ethanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/174/114/bd11ec7e-e4e1-481c-96bc-78099e87ad59/640/bd11ec7e-e4e1-481c-96bc-78099e87ad59.png)
2-[4-(2-Methylpropyl)phenyl]ethanol
![2-[4-(2-Methylpropyl)phenyl]ethanol certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/174/114/bd11ec7e-e4e1-481c-96bc-78099e87ad59/640/bd11ec7e-e4e1-481c-96bc-78099e87ad59.png)
Synonym(s):
4-(2-Methylpropyl)benzeneethanol, 4-Isobutylphenylethanol, NSC 69112
Empirical Formula (Hill Notation):
C12H18O
CAS Number:
Molecular Weight:
178.27
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing