
2,3,5-Trichlorobenzaldehyde

Synonym(s):
2,3,5-Trichlorobenzaldehyde
Empirical Formula (Hill Notation):
C7H3Cl3O
CAS Number:
Molecular Weight:
209.46
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing