Skip to Content
MilliporeSigma
1-Pentene analytical standard

1-Pentene

1-Pentene analytical standard
Linear Formula:
CH3CH2CH2CH=CH2
CAS Number:
Molecular Weight:
70.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing