
1-Methylcyclopropane-1-sulfonyl chloride

Empirical Formula (Hill Notation):
C4H7ClO2S
CAS Number:
Molecular Weight:
154.62
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing