
1-Acetoxy-1,3-butadiene

Synonym(s):
1,3-Butadienyl acetate
Linear Formula:
CH2=CHCH=CHOCOCH3
CAS Number:
Molecular Weight:
112.13
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1