Skip to Content
MilliporeSigma
1,3-Bis(dicyclohexylphosphino)propane bis(tetrafluoroborate) 97%

1,3-Bis(dicyclohexylphosphino)propane bis(tetrafluoroborate)

1,3-Bis(dicyclohexylphosphino)propane bis(tetrafluoroborate) 97%
Synonym(s):
dcpp
Linear Formula:
C27H50P2 · 2HBF4
CAS Number:
Molecular Weight:
612.26

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing