Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,3,5-TRIBROMO-2,4,6-TRIMETHYL-BENZENE
Linear Formula:
C
9
H
9
Br
3
CAS Number:
608-72-0
Molecular Weight:
356.884
Products
Formula Weight
Special Grade
Aldrich
CPR
(3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S97587
1,3,5-TRIBROMO-2,4,6-TRIMETHYL-BENZENE,
Aldrich
CPR
Expand
View Pricing
S109630
1,3,5-TRIBROMO-2,4,6-TRIMETHYL-BENZENE,
Aldrich
CPR
Expand
View Pricing
S869430
1,3,5-TRIBROMO-2,4,6-TRIMETHYL-BENZENE,
Aldrich
CPR
Expand
View Pricing