Skip to Content
MilliporeSigma
1,1,3,3,5,5,7,7,9,9,11,11,13,13,15,15-Hexadecamethyloctasiloxane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

1,1,3,3,5,5,7,7,9,9,11,11,13,13,15,15-Hexadecamethyloctasiloxane

1,1,3,3,5,5,7,7,9,9,11,11,13,13,15,15-Hexadecamethyloctasiloxane certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1,15-Dihydrohexadecamethyloctasiloxane
Empirical Formula (Hill Notation):
C16H50O7Si8
CAS Number:
Molecular Weight:
579.25

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1