10-O-Deacetyl-10-O-(3-oxobutanoyl)paclitaxel
Empirical Formula (Hill Notation):
C49H53NO15
Molecular Weight:
895.94
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing