Product Name
Lacto-N-tetraose from human milk, ≥95% (HPAE/PAD)
Product Number
l6770
Product Brand
sigma
CAS Number
14116-68-8
Molecular Weight
707.63
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 50MG/ML IN WATER
WATER BY KARL FISCHERNMT 15%
PROTON NMR SPECTRUMCONSISTENT WITH STRUCTURE
HPLC WITH PULSED AMPEROMETRIC DETECTIONMINIMUM 95%
DOCUMENT # L6770/04/02/99/1