Product Name
Atto 495 maleimide BioReagent, suitable for fluorescence
Product Number
41022
Product Brand
sigma
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Faint Red to Red
APPEARANCE (FORM)Powder
SOLUBILITY (COLOR)FOR INFORMATION
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)1 mg/ml in DMSO
DEGREE OF COUPLING≥ 90.0 %
EMISSION MAXIMUM524 ± 5 nm
REMARKS ON LUMINESCENCEExc. 500 nm in 0.1 M phosphate pH 7.0