Product Name
11-[5-(Dimethylamino)-1-naphthalenesulfonylamino]undecanoic acid BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Product Number
39235
Product Brand
sigma
CAS Number
73025-02-2
Molecular Weight
434.59
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (TLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EMISSION MAXIMUM516 ± 5 nm
REMARKS ON LUMINESCENCEEx: 335 nm in methanol