Product Name
1-(Trimethylacetyl)imidazole 98%
Product Number
660353
Product Brand
aldrich
CAS Number
4195-19-1
Molecular Weight
152.19
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER, CRYSTALS,
 CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID97.5% (MINIMUM)
 APPROVED APRIL 17, 2007 RJM