Product Name
Sodium 2-ethylhexanoate 97%
Product Number
511692
Product Brand
aldrich
CAS Number
19766-89-3
Molecular Weight
166.19
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Off White |
APPEARANCE (FORM) | Powder or Crystals or Chunk(s) |
PURITY (GC AREA %) | ≥ 96.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |