Product Name
Minimum Essential Medium Eagle With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Product Number
M2279
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance (Turbidity) | Clear |
Appearance (Form) | Solution |
pH | 7.0 - 7.6 |
Osmolality | 281 - 311 mOs/kg |
Cell Culture Testing - MTT | Pass |
Cell Line | Cell Line - Cell Types |
Sterility | Pass |
Endotoxin Level | <= 1 EU/ml |
Glucose Concentration | 0.9 - 1.1 g/l |