Product Name
2,5-Dichlorobenzoic acid PESTANAL®, analytical standard
Product Number
36705
Product Brand
SIAL
CAS Number
50-79-3
Molecular Weight
191.01
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 95.0 % |
IDENTIFICATION (LC-MS) | Identity Confirmed |
MELTING POINT | 150 - 156 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |