Product Name
2-Amino-5-methylhexane ≥99.0% (GC)
Product Number
08490
Product Brand
SIAL
CAS Number
28292-43-5
Molecular Weight
115.22
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow and Colorless to Light Green-Yellow |
APPEARANCE (FORM) | Liquid |
TITRATION (T) HCL 1M | 99.0 - 101.0 % |
PURITY (GC AREA %) | ≥ 99.0 % |
REFRACTIVE INDEX N20/D | 1.415 - 1.417 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |