Product Name
Ethyl laurate analytical standard
Product Number
07523
Product Brand
SIAL
CAS Number
106-33-2
Molecular Weight
228.37
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Almost Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 99.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |