Product Name
4,4′-Diaminostilbene dihydrochloride 95%
Product Number
D25206
Product Brand
ALDRICH
CAS Number
54760-75-7
Molecular Weight
283.20
Test | Specification |
---|---|
APPEARANCE (COLOR) | Off White to Light Brown |
APPEARANCE (FORM) | Powder |
ARGENTOMETR. TITRATION | 94.0 - 106.0 % |
PURITY (HPLC AREA %) | ≥ 94.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |